Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Qualification Standards Kit, MS, Liquid/Solid, System Performance

Catalog No. 50201695 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This is a Mass Spectrometry (Xevo TQ-S) standards kit designed for system assessment, used to conduct instrument calibration and benchmark the performance of the system. It contains 11 components.

  • Detection method: MS
  • Formulation: Liquid, Solid
  • Storage condition: 4°C (fridge)
  • System type: MS
  • Instrument: Xevo TQS
  • Shelf life: 12 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Type: Qualification
  • Application: System performance
  • Product type: Standard kits
  • Units per package: 1 pk
  • Components: 11 components, including various tuning solutions, set-up solutions, resolution solutions, calibration solutions, sensitivity standards, carryover test solutions, and mobile phase additives
  • Hazardous material: Contains hazardous material requiring special freight handling

Catalog No. 50-201-695 Supplier Waters Corp Supplier No. 700005472
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a Mass Spectrometry (Xevo TQ-S) standards kit designed for system assessment, used to conduct instrument calibration and benchmark the performance of the system. It contains 11 components.

  • Detection method: MS
  • Formulation: Liquid, Solid
  • Storage condition: 4°C (fridge)
  • System type: MS
  • Instrument: Xevo TQS
  • Shelf life: 12 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Type: Qualification
  • Application: System performance
  • Product type: Standard kits
  • Units per package: 1 pk
  • Components: 11 components, including various tuning solutions, set-up solutions, resolution solutions, calibration solutions, sensitivity standards, carryover test solutions, and mobile phase additives
  • Hazardous material: Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.