Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, Xevo TQ-S (Rotary), MS, with Chemical Kit
Catalog No. 50465915 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the Xevo TQ-S (rotary) Mass Spectrometer contains all the necessary parts to keep the instrument operating at peak performance on an annual basis. This kit also includes a chemical kit required to complete performance testing.
  • Performance maintenance for MS systems
  • Annual replacement schedule
  • Includes source, ESI probe, and vacuum pump components
  • Contains a chemical kit for performance testing
Catalog No. 50-465-915 Supplier Waters Corp Supplier No. 176002745
Add to Cart
Add to Cart
Request a Quote
This Performance Maintenance Kit for the Xevo TQ-S (rotary) Mass Spectrometer contains all the necessary parts to keep the instrument operating at peak performance on an annual basis. This kit also includes a chemical kit required to complete performance testing.
  • Performance maintenance for MS systems
  • Annual replacement schedule
  • Includes source, ESI probe, and vacuum pump components
  • Contains a chemical kit for performance testing