Learn More
Waters Corp Waters Performance Maintenance Kit, Xevo TQ-S (Rotary) Mass Spectrometer, Annual
Catalog No. 50466527 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is specifically designed for the Xevo TQ-S (rotary) Mass Spectrometer, providing all the essential quality parts required to maintain the instrument’s optimal performance annually.
  • Contains all quality parts necessary for annual instrument maintenance.
  • Ensures peak performance of the mass spectrometer.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
Catalog No. 50-466-527 Supplier Waters Corp Supplier No. 201000236
This Performance Maintenance Kit is specifically designed for the Xevo TQ-S (rotary) Mass Spectrometer, providing all the essential quality parts required to maintain the instrument’s optimal performance annually.
  • Contains all quality parts necessary for annual instrument maintenance.
  • Ensures peak performance of the mass spectrometer.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.