Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-XS MS System, Annual
Catalog No. 502392093 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed for the Xevo TQ-XS Mass Spectrometer to ensure annual peak performance. It contains essential quality parts, including source components and air filters. A chemical kit, necessary for completing performance testing, is not included with this specific kit.
  • Contains quality parts essential for the instrument's annual peak performance.
  • Includes source components.
  • Includes air filters.
  • Designed for Xevo TQ-XS Mass Spectrometer.
  • Has an annual replacement schedule.
  • A chemical kit for performance testing is not included.
Catalog No. 50-239-2093 Supplier Waters Corp Supplier No. 201000310
This Performance Maintenance Kit is designed for the Xevo TQ-XS Mass Spectrometer to ensure annual peak performance. It contains essential quality parts, including source components and air filters. A chemical kit, necessary for completing performance testing, is not included with this specific kit.
  • Contains quality parts essential for the instrument's annual peak performance.
  • Includes source components.
  • Includes air filters.
  • Designed for Xevo TQ-XS Mass Spectrometer.
  • Has an annual replacement schedule.
  • A chemical kit for performance testing is not included.