Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-XS Mass Spectrometer, Annual, Chemical Kit
Catalog No. 502391973 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit is designed for the Xevo TQ-XS Mass Spectrometer. It contains quality parts necessary to ensure the instrument operates at peak performance on an annual basis. This kit includes source components and air filters, and also provides a chemical kit essential for completing performance testing.
  • Contains quality parts for annual instrument maintenance.
  • Includes source components and air filters.
  • Chemical kit is required to complete performance testing.
  • Designed for MS system.
  • Recommended to order appropriate Tool Free Probe Capillary for the application.
Catalog No. 50-239-1973 Supplier Waters Corp Supplier No. 176004023
The Waters Performance Maintenance Kit is designed for the Xevo TQ-XS Mass Spectrometer. It contains quality parts necessary to ensure the instrument operates at peak performance on an annual basis. This kit includes source components and air filters, and also provides a chemical kit essential for completing performance testing.
  • Contains quality parts for annual instrument maintenance.
  • Includes source components and air filters.
  • Chemical kit is required to complete performance testing.
  • Designed for MS system.
  • Recommended to order appropriate Tool Free Probe Capillary for the application.