Learn More
Waters Corp Waters Corporation Xevo TQD/SQD2 (Rotary) Performance Maintenance Kit, Annual
Catalog No. 50466532 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit is designed to ensure Xevo TQD and SQD2 (rotary) Mass Spectrometers operate at peak performance annually. This kit contains essential components for the Source, ESI probe, and vacuum pump, providing all necessary Waters Quality Parts.<ul><li>Ensures instrument operates at peak performance.</li><li>Includes source components.</li><li>Includes ESI probe components.</li><li>Includes vacuum pump components.</li><li>Designed for annual replacement.</li><li>Compatible with Xevo TQD and SQD2 (rotary) mass spectrometer.</li></ul>
Catalog No. 50-466-532 Supplier Waters Corp Supplier No. 201000252
The Waters Performance Maintenance Kit is designed to ensure Xevo TQD and SQD2 (rotary) Mass Spectrometers operate at peak performance annually. This kit contains essential components for the Source, ESI probe, and vacuum pump, providing all necessary Waters Quality Parts.<ul><li>Ensures instrument operates at peak performance.</li><li>Includes source components.</li><li>Includes ESI probe components.</li><li>Includes vacuum pump components.</li><li>Designed for annual replacement.</li><li>Compatible with Xevo TQD and SQD2 (rotary) mass spectrometer.</li></ul>