Learn More
Waters Corp Performance Maintenance Kit, Xevo TQD/SQD 2 Scroll Pump, Annual
Catalog No. 50466533 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains all Waters Quality Parts™ needed to maintain the XEVO TQD and SQD2 (scroll) Mass Spectrometer for optimal annual performance. It includes Source components, ESI probe components, and vacuum pump components.
  • Source components
  • ESI probe components
  • Vacuum pump components
  • Performance maintenance type
  • For MS system type
  • Annual replacement schedule
  • One unit per package
  • Does not include the chemical kit required for performance testing
Catalog No. 50-466-533 Supplier Waters Corp Supplier No. 201000253
This kit contains all Waters Quality Parts™ needed to maintain the XEVO TQD and SQD2 (scroll) Mass Spectrometer for optimal annual performance. It includes Source components, ESI probe components, and vacuum pump components.
  • Source components
  • ESI probe components
  • Vacuum pump components
  • Performance maintenance type
  • For MS system type
  • Annual replacement schedule
  • One unit per package
  • Does not include the chemical kit required for performance testing