Learn More
Waters Corp Waters Performance Maintenance Kit, SQD2, Scroll Pump, Chemical Kit, Annual
Catalog No. 50465917 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the SQD 2 (Scroll) Mass Spectrometer is designed to keep the system operating at peak performance. This kit contains essential components for the source, ESI probe, and vacuum pump, and also includes a chemical kit necessary for completing performance testing.
  • Performance maintenance
  • Designed for MS system type
  • Annual replacement schedule
  • Includes source components, ESI probe components, and vacuum pump components
  • Contains a chemical kit for performance testing
  • One unit per package
Catalog No. 50-465-917 Supplier Waters Corp Supplier No. 176002781
The Waters Performance Maintenance Kit for the SQD 2 (Scroll) Mass Spectrometer is designed to keep the system operating at peak performance. This kit contains essential components for the source, ESI probe, and vacuum pump, and also includes a chemical kit necessary for completing performance testing.
  • Performance maintenance
  • Designed for MS system type
  • Annual replacement schedule
  • Includes source components, ESI probe components, and vacuum pump components
  • Contains a chemical kit for performance testing
  • One unit per package