Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies™ XF 1 M Glucose Solution, 50 mL

Catalog No. 103577100
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-357-7100 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-357-7100 Supplier Agilent Technologies™ Supplier No. 103577100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XF 1 M Glucose Solution specifically formulated for Agilent™ Seahorse XF assays to achieve optimal results.

  • For use with Seahorse XF24 Analyzer, Seahorse XFp Analyzer, Seahorse XFe24 Analyzer, and Seahorse XFe96 Analyzer
  • Refrigeration required at 2°C to 8°C

Specifications

Culture Type All
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile
Formulation Liquid with Glucose
For Use With (Application) Cell Culture and Cell-based Assays
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.