Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies™ XF 200 mM Glutamine Solution, 50 mL

Catalog No. 103579100
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-357-9100 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-357-9100 Supplier Agilent Technologies™ Supplier No. 103579100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XF 200 mM Glutamine Solution is specifically formulated for Agilent™ Seahorse XF assays to achieve optimal results.

  • For use with Seahorse XF24 Analyzer, Seahorse XFp Analyzer, Seahorse XFe24 Analyzer, and Seahorse XFe96 Analyzer
  • Freezer storage required at -20°C
  • Ships on dry ice

Specifications

Cell Line All
Cell Type All
Classification Animal Origin-free
Concentration 200 mM
Culture Type All
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-free
Sterility Sterile
With Additives None
Without Additives None
Formulation Liquid with Glutamine
Quantity 50 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.