Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies™ XF DMEM Medium, pH 7.4

Catalog No. 103575100
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-357-5100 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-357-5100 Supplier Agilent Technologies™ Supplier No. 103575100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XF DMEM Medium, pH 7.4 is a DMEM-based medium validated for use with the Seahorse Real-Time ATP rate assay and the glycolytic rate assay. It is also recommended to use for all Seahorse XF assays.

  • Contains no phenol red, bicarbonate, glucose, glutamine, or pyruvate, allowing for full customization
  • XF media with pH 7.4 is also available, simplifying the workflow, saving time for assay preparation, and ensuring consistent results across experiments
  • Contains 5 mM HEPES, and the pH of this medium is pre-adjusted to 7.4
  • To ensure a proper final pH in assay media, Seahorse XF supplements must be used at recommended concentration range

Specifications

Cell Line All
Cell Type All
Classification Animal Origin-free
Concentration 1X
Form Liquid
Product Type Seahorse XF Assay
Serum Level Serum-free
Sterility Sterile
With Additives None
Without Additives None
Quantity 500 mL
Formulation Liquid XF Medium Assay
pH 7.4
Culture Type Adherent Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.