Learn More
Agilent Technologies™ XF HS Miniplates Set
Description
- Available in a wide range of configurations to accomodate 2D monolayers, 3D spheroids, suspension cells, islets, and isolated mitochondria
- Conforms to ANSI/SLAS Microplate Standards for footprint dimensions and well positions
- Tissue culture treated for optimum adherence and growth of mammalian cells
- Gamma-irradiated for use with live cell culture
- Molded stops prevent the sensor cartridge sleeves from disrupting your cell monolayer
- XF HS miniplate (XF HS Mini analyzer compatible) enables 1/3 fewer cells per well
- Pre-coated Poly-D-Lysine cell culture plates, available in 96-well and 8-well formats, are available for use with XF assays using suspension cell, providing convenience and consistent assay result
- PDL microplates are manufactured in an aseptic facility
- Designed specifically for use with Seahorse XF Analyzers
Specifications
Specifications
| Barcode | Includes Barcode |
| Array | 1 x 8 |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Equipment) | Seahorse XF HS Mini Analyzer |
| Material | Polystyrene |
| No. of Wells | 8 |
| Sterility | Sterile |
| Surface Treatment | Plasma |
| Well Shape | Flat Bottom (F-well) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.