Learn More
Agilent Technologies Seahorse XF24 V7 PS Cell Culture Microplates. For use with both Seahorse XFe24 and XF24 Analyzers,XF cell culture microplates are tissue culture treated for optimum adherence and growth of mammalian cells. The XF microplates are supplied in individual gamma irradiated containers. 10/pk