Learn More
Aat Bioquest XFD532 PEG4 biotin conjugate, 1mg
SDP
Catalog No. NC3919077
Encompass_Preferred
The XFD532 PEG4 biotin conjugate incorporates the XFD532 fluorophore, which is structurally identical to Alexa Fluor 532 and efficiently excited at 532 nm. It exhibits strong orange fluorescence and high avidin-binding efficiency, closely resembling native biotin in affinity, rapid association, and non-cooperative binding to avidin and streptavidin tetramers. This conjugate is particularly well suited for the detection and quantification of biotin-binding sites on avidin, streptavidin, or neutravidin. In contrast to Biotin-4-fluorescein, which suffers from poor aqueous solubility, pH-dependent absorbance, and short-wavelength emission, the XFD532 PEG4 biotin conjugate is fully water soluble and maintains pH-independent fluorescence across the range of pH 4–10.
Catalog No. NC3919077 Supplier Aat Bioquest Supplier No. 3110
The XFD532 PEG4 biotin conjugate incorporates the XFD532 fluorophore, which is structurally identical to Alexa Fluor 532 and efficiently excited at 532 nm. It exhibits strong orange fluorescence and high avidin-binding efficiency, closely resembling native biotin in affinity, rapid association, and non-cooperative binding to avidin and streptavidin tetramers. This conjugate is particularly well suited for the detection and quantification of biotin-binding sites on avidin, streptavidin, or neutravidin. In contrast to Biotin-4-fluorescein, which suffers from poor aqueous solubility, pH-dependent absorbance, and short-wavelength emission, the XFD532 PEG4 biotin conjugate is fully water soluble and maintains pH-independent fluorescence across the range of pH 4–10.