Learn More
Abcam x FD750 Conjugation Kit, 100UG
Catalog No. 502700604
Encompass_Preferred
XFD750 conjugation / labeling in < 4 hrs with 30 secs hands-on time using XFD750 Conjugation Kit ab312814. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-0604 Supplier Abcam Supplier No. AB312814100UG
XFD750 conjugation / labeling in < 4 hrs with 30 secs hands-on time using XFD750 Conjugation Kit ab312814. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement