Learn More
Agilent Technologies XFE96/XF PRO PDL FLUXPAK MINI
Catalog No. NC2459380
Encompass
Seahorse XFe96/XF Pro PDL FluxPak Mini. For use with XFe96 and XF Pro Analyzer. Seahorse XFe96/XF Pro PDL FluxPak Mini includes 6 XFe96/XF Pro sensor cartridges, 6 XFe96/XF Pro PDL Cell Culture Microplates, and 1 bottle of Seahorse XF Calibrant Solution 500mL.
Catalog No. NC2459380 Supplier Agilent Technologies Supplier No. 103798100
Seahorse XFe96/XF Pro PDL FluxPak Mini. For use with XFe96 and XF Pro Analyzer. Seahorse XFe96/XF Pro PDL FluxPak Mini includes 6 XFe96/XF Pro sensor cartridges, 6 XFe96/XF Pro PDL Cell Culture Microplates, and 1 bottle of Seahorse XF Calibrant Solution 500mL.