Learn More
Takara Bio Xfect™ RNA Transfection Reagent | 1.2mL
SDP
Catalog No. NC1102016
Encompass
Xfect RNA Transfection Reagent is a complete system for highly efficient transfection of mammalian cells with all types of RNA, including messenger RNA (mRNA), small interfering RNA (siRNA), and single guide RNA (sgRNA).The protocol is simple, and transfections can be carried out entirely in the presence of serum. Xfect RNA Transfection Reagent creates biodegradable nanoparticles when combined with RNA, resulting in very low cytotoxicity.
Catalog No. NC1102016 Supplier Takara Bio Supplier No. 631450
Xfect RNA Transfection Reagent is a complete system for highly efficient transfection of mammalian cells with all types of RNA, including messenger RNA (mRNA), small interfering RNA (siRNA), and single guide RNA (sgRNA).The protocol is simple, and transfections can be carried out entirely in the presence of serum. Xfect RNA Transfection Reagent creates biodegradable nanoparticles when combined with RNA, resulting in very low cytotoxicity.