Learn More
Takara Bio Xfect™ Transfection Reagent 300 Rxns
SDP
Catalog No. NC0147818
Encompass
Xfect is a transfection reagent that creates biodegradable nanoparticles that permit superior transfection efficiency of plasmid DNA into mammalian cells. Transfections can be carried out entirely in the presence of serum. 300 Rxns
Catalog No. NC0147818 Supplier Takara Bio Supplier No. 631318
Xfect is a transfection reagent that creates biodegradable nanoparticles that permit superior transfection efficiency of plasmid DNA into mammalian cells. Transfections can be carried out entirely in the presence of serum. 300 Rxns