Learn More
Avantor Fluid Handling Avantor Fluid Handling Transfer Tubing Assembly, Gamma Irradiated, Animal-Derived Component Free, 50 Units
Catalog No. NC2680072
Encompass
The Avantor Fluid Handling XFER Tubing Assembly is a reliable single-use fluid transfer solution ideal for scientific and bioprocessing applications. Manufactured under strict standards, it features gamma irradiation for optimal sterility (SAL 10^(-6)) and is compliant with USP Class VI for biocompatibility. Its animal-derived component free (ADCF) materials ensure high safety levels. Conveniently packaged single-bagged, this assembly stands out for its high sterility assurance, making it an essential choice for critical fluid pathways.
Catalog No. NC2680072 Supplier Avantor Fluid Handling Supplier No. ICITA12269
The Avantor Fluid Handling XFER Tubing Assembly is a reliable single-use fluid transfer solution ideal for scientific and bioprocessing applications. Manufactured under strict standards, it features gamma irradiation for optimal sterility (SAL 10^(-6)) and is compliant with USP Class VI for biocompatibility. Its animal-derived component free (ADCF) materials ensure high safety levels. Conveniently packaged single-bagged, this assembly stands out for its high sterility assurance, making it an essential choice for critical fluid pathways.