Learn More
Agilent Technologies™ XFp PDL Miniplates
Description
- Available in a wide range of configurations to accommodate 2D monolayers, 3D spheroids, suspension cells, islets, and isolated mitochondria
- Solid-state sensor probes containing polymer embedded fluorophores detect oxygen and proton levels in cell culture media
- Patented integrated injection ports for adding compounds, stimulators, inhibitors, substrates, and beads conjugated with antigens
- Patented transient microchamber provides increased analytical sensitivity and signal to noise of OCR and ECAR (PER, gPER) readings when used in an XF Analyzer
- Sensors are spotted onto the cartridge using a multi-step process that includes manual and machine vision inspection
- Conforms to ANSI/SLAS Microplate Standards for footprint dimensions and well positions
- Pre-coated Poly-D-Lysine cell culture plates, available in 96-well and 8-well formats, are available for use with XF assays using suspension cell, providing convenience and consistent assay result
- PDL cell culture plates are manufactured in an aseptic facility
- XF Pro M cell culture plates are now available for use with XF Pro analyzer to minimize assay edge effect and improve intra-plate consistency
- Designed specifically for use with Seahorse XF Analyzers
Specifications
Specifications
| Barcode | Includes Barcode |
| Array | 1 x 8 |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Equipment) | Seahorse XF HS Mini Analyzer, Seahorse XFp Analyzer |
| Material | Polystyrene |
| No. of Wells | 8 |
| Sterility | Non-sterile |
| Surface Treatment | Poly-D-Lysine |
| Well Shape | Flat Bottom (F-well) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.