Learn More
Medchemexpress LLC XG/Xg blood group H 50ug
SDP
Catalog No. 5000180065
Encompass_Preferred
XG/Xg blood group Protein is a type I transmembrane glycoprotein belonging to the CD99 family It is a erythrocyte membrane protein that is used as the basis of the Xg blood group system XG/Xg blood group Protein Human (HEK293 His) is the recombinant human-derived XG/Xg blood group protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80065 Supplier Medchemexpress LLC Supplier No. HYP7353850UG
XG/Xg blood group Protein is a type I transmembrane glycoprotein belonging to the CD99 family It is a erythrocyte membrane protein that is used as the basis of the Xg blood group system XG/Xg blood group Protein Human (HEK293 His) is the recombinant human-derived XG/Xg blood group protein expressed by HEK293 with C-His labeled tag