Learn More
MSC Makita 40V XGT Dust Extractor, 1/2 gal. HEPA, 1 ea
Catalog No. 502817322
Encompass
This portable and powerful cordless dry dust extractor kit offers a clean-up solution for construction and cleaning applications, bridging the gap between small on-board systems and larger, less portable wheeled vacuums. It is designed for efficient dust extraction with ultimate portability.
  • Generates 67 CFM suction and 72 in static water lift.
  • Auto-start Wireless System (AWS®) capable for tool-to-extractor communication.
  • Up to one hour run time on medium with 40V XGT® 5.0Ah battery.
  • Efficient two-stage HEPA filtration (captures 99.97% of 0.3 micron particulates).
  • Offers three-speed power selection.
  • Padded shoulder straps and waist belt for comfortable fit.
  • Equipped with anti-static hose and adapter cuffs.
Catalog No. 50-281-7322 Supplier MSC Supplier No. 06301675
This portable and powerful cordless dry dust extractor kit offers a clean-up solution for construction and cleaning applications, bridging the gap between small on-board systems and larger, less portable wheeled vacuums. It is designed for efficient dust extraction with ultimate portability.
  • Generates 67 CFM suction and 72 in static water lift.
  • Auto-start Wireless System (AWS®) capable for tool-to-extractor communication.
  • Up to one hour run time on medium with 40V XGT® 5.0Ah battery.
  • Efficient two-stage HEPA filtration (captures 99.97% of 0.3 micron particulates).
  • Offers three-speed power selection.
  • Padded shoulder straps and waist belt for comfortable fit.
  • Equipped with anti-static hose and adapter cuffs.