Learn More
MSC Makita Backpack Dry Vacuum Kit, 40V max XGT Brushless, 2 Quart HEPA, 5.0Ah
Catalog No. 502837714
Encompass
This 40V max XGT Brushless Cordless 2 Quart HEPA Filter Backpack Dry Vacuum Kit offers a powerful, portable, and cord-free cleaning solution. It is designed for cleaning crews and users needing a compact, efficient, and comfortable vacuum for various applications.
  • Up to 85 CFM suction power and 72 inch static water lift.
  • Two-stage HEPA filtration captures 99.97% of particulates 0.3 microns and larger.
  • Up to 1 hour run time on medium setting with 5.0Ah battery.
  • Compact 2 quart size for portability and cleaning in confined spaces.
  • Features 4-speed power selection for run time management.
  • Padded shoulder straps and waist belt ensure user comfort.
  • Includes anti-static hose, telescoping wand, and floor nozzle.
Catalog No. 50-283-7714 Supplier MSC Supplier No. 41962291
This 40V max XGT Brushless Cordless 2 Quart HEPA Filter Backpack Dry Vacuum Kit offers a powerful, portable, and cord-free cleaning solution. It is designed for cleaning crews and users needing a compact, efficient, and comfortable vacuum for various applications.
  • Up to 85 CFM suction power and 72 inch static water lift.
  • Two-stage HEPA filtration captures 99.97% of particulates 0.3 microns and larger.
  • Up to 1 hour run time on medium setting with 5.0Ah battery.
  • Compact 2 quart size for portability and cleaning in confined spaces.
  • Features 4-speed power selection for run time management.
  • Padded shoulder straps and waist belt ensure user comfort.
  • Includes anti-static hose, telescoping wand, and floor nozzle.