Learn More
New England Biolabs, Inc. XhoI – 25000 units
SDP
Catalog No. 50994935
Encompass
A restriction endonuclease that recognizes the sequence C^TCGA_G.
Catalog No. 50-994-935 Supplier New England Biolabs, Inc. Supplier No. R0146M
A restriction endonuclease that recognizes the sequence C^TCGA_G.