Learn More
New England Biolabs, Inc. XhoI – 5000 units
SDP
Catalog No. 50812025
Encompass
A restriction endonuclease that recognizes the sequence C^TCGA_G.
Catalog No. 50-812-025 Supplier New England Biolabs, Inc. Supplier No. R0146S
A restriction endonuclease that recognizes the sequence C^TCGA_G.