Learn More
Ellsworth Adhesive Co XIAMETER PMX-200 10CS 1GL
Catalog No. NC1785065
Encompass
Dow XIAMETER™ PMX-200 Silicone Fluid, 10 CS is a polydimethyl-siloxane based fluid that is used for industrial applications, electrical insulating, penetrating oils, coating, and used in personal care products such as lotions, deodorants, nail polish, and hair spray. It offers high shearability, spreadability, compressibility, and low vapor pressure, reactivity, and surface tension. It has good dielectric properties, heat stability, water resistance, and leveling.
Catalog No. NC1785065 Supplier Ellsworth Adhesive Co Supplier No. PMX200 10CSGL
Dow XIAMETER™ PMX-200 Silicone Fluid, 10 CS is a polydimethyl-siloxane based fluid that is used for industrial applications, electrical insulating, penetrating oils, coating, and used in personal care products such as lotions, deodorants, nail polish, and hair spray. It offers high shearability, spreadability, compressibility, and low vapor pressure, reactivity, and surface tension. It has good dielectric properties, heat stability, water resistance, and leveling.