Learn More
Medchemexpress LLC XIAP degrader-1 5mg | 2803617-91-4 | 587.75 | C34H45N5O4 | 5 MG
SDP
Catalog No. 5000227956
Encompass_Preferred
XIAP degrader-1 is a primary amine-tethered small molecule that promotes degradation of X-linked inhibitor of apoptosis protein (XIAP). It is supplied for research use and is intended for biochemical and cellular studies of IAP-mediated apoptosis.
  • Primary amine-tethered small molecule that promotes XIAP degradation.
  • Useful for biochemical and cellular studies of IAP-mediated apoptosis.
  • High purity (>98%), suitable for research applications.
  • Available in 1 mg, 5 mg, and 10 mg sizes for small-scale experiments.
Catalog No. 50-002-27956 Supplier Medchemexpress LLC Supplier No. HY1158655MG
XIAP degrader-1 is a primary amine-tethered small molecule that promotes degradation of X-linked inhibitor of apoptosis protein (XIAP). It is supplied for research use and is intended for biochemical and cellular studies of IAP-mediated apoptosis.
  • Primary amine-tethered small molecule that promotes XIAP degradation.
  • Useful for biochemical and cellular studies of IAP-mediated apoptosis.
  • High purity (>98%), suitable for research applications.
  • Available in 1 mg, 5 mg, and 10 mg sizes for small-scale experiments.