Learn More
Medchemexpress LLC HY-10787 1mg Medchemexpress, Ximelagatran CAS:192939-46-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107871MG
Encompass_Preferred
Medchemexpress, HY-10787 1mg Ximelagatran CAS:192939-46-1 Ximelagatran (H 376/95) is an orally active thrombin inhibitor that selectively and competitively inhibits both free and clot-bound thrombin. Ximelagatran is an anticoagulant agent with a rapid onset of anticoagulant effect, predictable, dose-dependent pharmcokinetics and pharmacodynamics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8477