Learn More
Medchemexpress LLC Ximelagatran 500ug | 192939-46-1 | 500UG
SDP
Catalog No. 5000242301
Encompass_Preferred
Ximelagatran (H 376/95) is an orally active thrombin inhibitor that selectively and competitively inhibits both free and clot-bound thrombin Ximelagatran is an anticoagulant agent with a rapid onset of anticoagulant effect predictable dose-dependent pharmcokinetics and pharmacodynamics[1 [2
Catalog No. 50-002-42301 Supplier Medchemexpress LLC Supplier No. HY10787500UG
Ximelagatran (H 376/95) is an orally active thrombin inhibitor that selectively and competitively inhibits both free and clot-bound thrombin Ximelagatran is an anticoagulant agent with a rapid onset of anticoagulant effect predictable dose-dependent pharmcokinetics and pharmacodynamics[1 [2