Learn More
Cayman Chemical Ximelagatran, Purity: ≥98%, CAS: 192939-46-1, 10 mg
SDP
Catalog No. 502980293
Encompass_Preferred
An ester prodrug of melagatran, a potent, direct, and reversible thrombin inhibitor (Ki = 1.2 nM); used as an anticoagulant in a variety of situations
Catalog No. 50-298-0293 Supplier Cayman Chemical Supplier No. 1686210 MG
An ester prodrug of melagatran, a potent, direct, and reversible thrombin inhibitor (Ki = 1.2 nM); used as an anticoagulant in a variety of situations