Learn More
Sigma Aldrich Xion Aem-pention-72-15cl Thickness 5um L X W 5cmx5cm
Catalog No. AEM2B0522 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
AEM2B0522 1 Ea.
1 options
Catalog No. AEM2B0522 Supplier Sigma Aldrich Supplier No. AEM2B05221EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
XION Composite PENTION Membranes are ultra-thin, ultra-strong, and provide ultra-high performance while demonstrating highly stable performance as reinforced anion exchange membranes (AEM). The ionomer structure contains a poly(norbornene) backbone with quaternary ammonium functional groups. A reinforcement layer is integrated into the structure of the membrane to provide enhanced mechanical properties. The enhanced mechanical properties as free-standing membranes, providing higher ionic conductance without sacrificing strength. Applications for XION Composite PENTION Membranes include fuel cells, electrolysis, and flow batteries.Product of Xergy
Specifications
Quantity 1 Ea.