Learn More
Medchemexpress LLC HY-W042301 50mg Medchemexpress, Xipamide CAS:14293-44-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW04230150MG
Encompass_Preferred
Medchemexpress, HY-W042301 50mg Xipamide CAS:14293-44-8 Xipamide is a sulfonamide-based diuretic. Xipamide is an antihypertensive agent able to selectively inhibit the anion exchanger (AE). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3480