Learn More
Medchemexpress LLC XJB-5-131 | 866404-31-1 | 99.9% | 1 MG
SDP
Catalog No. 5000341320
Encompass_Preferred
XJB-5-131 is a synthetic, mitochondria-targeted antioxidant that acts as a bi-functional ROS and electron scavenger. It functions as an effective ionizing irradiation protector and mitigator for cord blood mononuclear cells.
  • Ameliorates hemorrhagic shock (HS)-induced activation of pro-apoptotic enzymes.
  • Reduces apoptosis and enhances cell survival.
  • Functions as a radiation protector for CFU-GM.
  • Ameliorates peroxidation of mitochondrial phospholipid, cardiolipin.
  • Prolongs survival in rats after profound blood loss.
  • Reduces oxidative damage to mitochondrial DNA and improves mitochondrial function in Huntington's disease models.
Catalog No. 50-003-41320 Supplier Medchemexpress LLC Supplier No. HY1294601MG
XJB-5-131 is a synthetic, mitochondria-targeted antioxidant that acts as a bi-functional ROS and electron scavenger. It functions as an effective ionizing irradiation protector and mitigator for cord blood mononuclear cells.
  • Ameliorates hemorrhagic shock (HS)-induced activation of pro-apoptotic enzymes.
  • Reduces apoptosis and enhances cell survival.
  • Functions as a radiation protector for CFU-GM.
  • Ameliorates peroxidation of mitochondrial phospholipid, cardiolipin.
  • Prolongs survival in rats after profound blood loss.
  • Reduces oxidative damage to mitochondrial DNA and improves mitochondrial function in Huntington's disease models.