Learn More
Sigma Aldrich Fine Chemicals Biosciences XK 26 00 Column
Catalog No. 502908757
Encompass
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to A„KTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous SOLUTIONons and most organic solvents used in liquid chromatography of macromolecules.
Catalog No. 50-290-8757 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28988951
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to A„KTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous SOLUTIONons and most organic solvents used in liquid chromatography of macromolecules.