Learn More
Aviva Systems Biology Corp XK Antibody - N-terminal region (ARP33808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human XK. Alternate names: KX, X1k, XKR1, MCLDSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55154