Learn More
ABclonal Technology XK Rabbit pAb
SDP
Catalog No. 5000106416
Encompass_Preferred
This locus controls the synthesis of the Kell blood group precursor substance (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. The encoded protein has structural characteristics of prokaryotic and eukaryotic membrane transport proteins.
Catalog No. 50-001-06416 Supplier ABclonal Technology Supplier No. A151091000UL
This locus controls the synthesis of the Kell blood group precursor substance (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. The encoded protein has structural characteristics of prokaryotic and eukaryotic membrane transport proteins.