Learn More
Miele, Inc. XKM RS232 10 Med Serial Module
Catalog No. 502870742
Encompass_Preferred
XKM RS232 10 Med Serial Module for connection to PRT Printer, suitable for all PG85 Flex
Catalog No. 50-287-0742 Supplier Miele, Inc. Supplier No. 10444420
XKM RS232 10 Med Serial Module for connection to PRT Printer, suitable for all PG85 Flex