Learn More
Selleck Chemical LLC Cabozantinib (BMS-907351) 200mg 849217-68-1 XL184
SDP
Catalog No. 506227
Encompass_Preferred
Cabozantinib (XL184, BMS-907351) is a potent VEGFR2 inhibitor with IC50 of 0.035 nM and also inhibits c-Met, Ret, Kit, Flt-1/3/4, Tie2, and AXL with IC50 of 1.3 nM, 4 nM, 4.6 nM, 12 nM/11.3 nM/6 nM, 14.3 nM and 7 nM in cell-free assays, respectively. Cabozantinib induces PUMA-dependent apoptosis in colon cancer cells via AKT/GSK-3?/NF-?B signaling pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-622-7 Supplier Selleck Chemical LLC Supplier No. S1119200MG
Cabozantinib (XL184, BMS-907351) is a potent VEGFR2 inhibitor with IC50 of 0.035 nM and also inhibits c-Met, Ret, Kit, Flt-1/3/4, Tie2, and AXL with IC50 of 1.3 nM, 4 nM, 4.6 nM, 12 nM/11.3 nM/6 nM, 14.3 nM and 7 nM in cell-free assays, respectively. Cabozantinib induces PUMA-dependent apoptosis in colon cancer cells via AKT/GSK-3?/NF-?B signaling pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.