Learn More
MilliporeSigma XL 300 MILLIPORE EXPRESS OPTI
Catalog No. NC1537617
Encompass
Opticap Gamma Compatible XL 300 Millipore Express SHC 0.5/0.2 m 3/4 in. TC-9/16 in. HB with Bell
Catalog No. NC1537617 Supplier MilliporeSigma Supplier No. KHGEG003FB3
Opticap Gamma Compatible XL 300 Millipore Express SHC 0.5/0.2 m 3/4 in. TC-9/16 in. HB with Bell