Learn More
Medchemexpress LLC XL-3158 | 3117797-77-7 | 99.5% | 690.61 | 200 MG
SDP
Catalog No. 5000386877
Encompass_Preferred
XL-3158 is a selective and cross-species Cyclic GMP-AMP synthase (cGAS) inhibitor. It simultaneously occupies allosteric and orthosteric sites, stabilizing the activation loop in a closed, inactive conformation, thereby attenuating cGAS-DNA interactions. It efficiently penetrates cells by inhibiting aggregate formation, effectively reducing the local concentration of cGAS within cells. This compound can be used for the study of cGAS-dependent inflammatory diseases.
  • Blocks cGAS intracellular condensation with DNA by inhibiting cGAS phase separation in L929 cells.
  • Specifically inhibits downstream signaling activation of the cGAS-STING pathway in THP-1 Dual cells.
  • Selectively inhibits the DNA-triggered cGAS-dependent interferon pathway in THP-1 Dual cells.
  • Has CC50 values of 77.55 μM against THP-1 cells and 56.79 μM against RAW-ISG cells.
  • Attenuates Cerulein-induced acute pancreatitis.
Catalog No. 50-003-86877 Supplier Medchemexpress LLC Supplier No. HY174802200MG
XL-3158 is a selective and cross-species Cyclic GMP-AMP synthase (cGAS) inhibitor. It simultaneously occupies allosteric and orthosteric sites, stabilizing the activation loop in a closed, inactive conformation, thereby attenuating cGAS-DNA interactions. It efficiently penetrates cells by inhibiting aggregate formation, effectively reducing the local concentration of cGAS within cells. This compound can be used for the study of cGAS-dependent inflammatory diseases.
  • Blocks cGAS intracellular condensation with DNA by inhibiting cGAS phase separation in L929 cells.
  • Specifically inhibits downstream signaling activation of the cGAS-STING pathway in THP-1 Dual cells.
  • Selectively inhibits the DNA-triggered cGAS-dependent interferon pathway in THP-1 Dual cells.
  • Has CC50 values of 77.55 μM against THP-1 cells and 56.79 μM against RAW-ISG cells.
  • Attenuates Cerulein-induced acute pancreatitis.