Learn More
Medchemexpress LLC XL-3158 | 99.6% | 690.61 g/mol | C30H17F7N6O2S2 | 10 MG
SDP
Catalog No. 5000299132
Encompass_Preferred
XL-3158 is a selective, cross-species inhibitor of cyclic GMP-AMP synthase (cGAS) for research applications. It binds both allosteric and orthosteric sites to stabilize the activation loop in an inactive conformation, reducing cGAS-DNA interactions and phase separation. The compound shows cellular activity with limited cytotoxicity at effective concentrations and is suitable for studies of cGAS-dependent inflammatory pathways.
  • Selective cross-species inhibition of cGAS
  • Dual-site binding stabilizes inactive activation loop
  • Reduces cGAS-DNA interactions and phase separation
  • Demonstrated cellular activity with low cytotoxicity
  • High purity suitable for cell-based assays
Catalog No. 50-002-99132 Supplier Medchemexpress LLC Supplier No. HY17480210MG
XL-3158 is a selective, cross-species inhibitor of cyclic GMP-AMP synthase (cGAS) for research applications. It binds both allosteric and orthosteric sites to stabilize the activation loop in an inactive conformation, reducing cGAS-DNA interactions and phase separation. The compound shows cellular activity with limited cytotoxicity at effective concentrations and is suitable for studies of cGAS-dependent inflammatory pathways.
  • Selective cross-species inhibition of cGAS
  • Dual-site binding stabilizes inactive activation loop
  • Reduces cGAS-DNA interactions and phase separation
  • Demonstrated cellular activity with low cytotoxicity
  • High purity suitable for cell-based assays