Learn More
Apexbio Technology LLC XL-888 1149705-71-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061766
Encompass_Preferred
XL-888 (CAS 1149705-71-4) is an orally bioavailable small molecule inhibitor targeting heat shock protein 90 (HSP90) It selectively inhibits HSP90 and HSP90 with IC50 values of 22 nM and 44 nM respectively while exhibiting markedly weaker inhibition against a panel of other kinases Structural analyses indicated XL-888 binds HSP90 via hydrogen bonding interactions involving the Asp93 residue In tumor cell assays XL-888 potently suppresses proliferation with IC50 values in the range of 0 1-45 5 nM XL-888 is used in oncology research particularly for studying resistance mechanisms to targeted therapies
Catalog No. 50-000-61766 Supplier Apexbio Technology LLC Supplier No. A43885.1
XL-888 (CAS 1149705-71-4) is an orally bioavailable small molecule inhibitor targeting heat shock protein 90 (HSP90) It selectively inhibits HSP90 and HSP90 with IC50 values of 22 nM and 44 nM respectively while exhibiting markedly weaker inhibition against a panel of other kinases Structural analyses indicated XL-888 binds HSP90 via hydrogen bonding interactions involving the Asp93 residue In tumor cell assays XL-888 potently suppresses proliferation with IC50 values in the range of 0 1-45 5 nM XL-888 is used in oncology research particularly for studying resistance mechanisms to targeted therapies