Learn More
Cole-Parmer Nylon Bottle/Cylinder Brush, 18" Handle x 6" L x 2.375" Diameter; 12/Pk
Catalog No. NC0316471 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.
Catalog No. NC0316471 Supplier Cole-Parmer Supplier No. 8455018
Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.