Learn More
Cole-Parmer Nylon Bottle/Cylinder Brush, 24" Handle x 6" L x 2.75" Diameter; 12/Pk
Catalog No. NC0905031 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.
Catalog No. NC0905031 Supplier Cole-Parmer Supplier No. 8455019
Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.