Learn More
Quality Lab Accessories XL CAPSULE SINKER 3.4 X 1.3CM
Catalog No. NC9383171
Encompass
Spiral Coated Capsule Sinker, PTFE Coated Music Wire, 1.15" (29.2mm) L x .465" (11.8mm) W capacity, 5.5 coils
Catalog No. NC9383171 Supplier Quality Lab Accessories Supplier No. CAPWHTXL
Spiral Coated Capsule Sinker, PTFE Coated Music Wire, 1.15" (29.2mm) L x .465" (11.8mm) W capacity, 5.5 coils