Learn More
Metasystems Group Inc XL-CDKN2C/CKS1B Enumeration Probe, 100 μL, Green and Orange, Fluorescence in situ hybridization (FISH), Copy number variation detection, Clinical indication: Multiple Myeloma
SDP
Catalog No. NC1020116
Encompass
The XL CDKN2C/CKS1B enumeration probe is a fluorescence in situ hybridization (FISH) test for the detection of copy number variations in the CDKN2C and CKS1B gene regions. It is designed as a diagnostic aid for disease monitoring in patients with multiple myeloma (MM). The probe uses a green label for the CDKN2C gene at 1p32.3 and an orange label for the CKS1B gene at 1q21-22. This ensures robust detection of deletions or amplifications on chromosome 1, providing reliable clinical results. The product is IVDR-certified under Regulation (EU) 2017/746.
  • Detection of copy number variations
  • Low background and consistent signal strength
  • Designed for multiple myeloma applications
  • Certified for in vitro diagnostic use
  • Includes 100 μL for 10 tests
Catalog No. NC1020116 Supplier Metasystems Group Inc Supplier No. D5099999OG
Add to Cart
Add to Cart
Request a Quote
The XL CDKN2C/CKS1B enumeration probe is a fluorescence in situ hybridization (FISH) test for the detection of copy number variations in the CDKN2C and CKS1B gene regions. It is designed as a diagnostic aid for disease monitoring in patients with multiple myeloma (MM). The probe uses a green label for the CDKN2C gene at 1p32.3 and an orange label for the CKS1B gene at 1q21-22. This ensures robust detection of deletions or amplifications on chromosome 1, providing reliable clinical results. The product is IVDR-certified under Regulation (EU) 2017/746.
  • Detection of copy number variations
  • Low background and consistent signal strength
  • Designed for multiple myeloma applications
  • Certified for in vitro diagnostic use
  • Includes 100 μL for 10 tests