Learn More
TWD Scientific LLC Extra large Polypropylene Weighing Funnels. Pack of 10
SDP
Catalog No. 501173374 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable polypropylene weighing funnel, size extra large. Fits volumetric flask volumes (mL) of 1000, 2000. Funnels have a maximum capacity of 60mL Solid/25mL Liquid. 10/Pack
Catalog No. 50-117-3374 Supplier TWD Scientific LLC Supplier No. DPWFPP4XLPK
Disposable polypropylene weighing funnel, size extra large. Fits volumetric flask volumes (mL) of 1000, 2000. Funnels have a maximum capacity of 60mL Solid/25mL Liquid. 10/Pack