Learn More
Metasystems Group Inc XL DLEU/LAMP probe, 10 x 100 μL, orange-labeled fluorophore, green-labeled probe
SDP
Catalog No. NC1020117
Encompass
The METASYSTEMS GROUP INC probe is a non-automated fluorescence in situ hybridization (FISH) tool specifically designed to detect deletions in the gene region DLEU1/MIR15A/MIR16-1 at 13q14.2, playing a vital role in diagnosing certain hematological malignancies like chronic lymphocytic leukemia. This probe features both orange and green labels to enhance detection accuracy and is IVDR-certified.
  • High diagnostic sensitivity of 99.3% for DLEU1 deletions
  • Diagnostic specificity of 100% for DLEU1 deletions
  • Designed for use in chronic lymphocytic leukemia, multiple myeloma, and non-Hodgkin-lymphomas
  • Non-automated qualitative test method
  • Utilizes methanol/acetic-acid fixed cells from various sample types
  • Regulatory status: IVDR certified
  • 10 x 100 μL package configuration
Catalog No. NC1020117 Supplier Metasystems Group Inc Supplier No. D5054999OG
Add to Cart
Add to Cart
Request a Quote
The METASYSTEMS GROUP INC probe is a non-automated fluorescence in situ hybridization (FISH) tool specifically designed to detect deletions in the gene region DLEU1/MIR15A/MIR16-1 at 13q14.2, playing a vital role in diagnosing certain hematological malignancies like chronic lymphocytic leukemia. This probe features both orange and green labels to enhance detection accuracy and is IVDR-certified.
  • High diagnostic sensitivity of 99.3% for DLEU1 deletions
  • Diagnostic specificity of 100% for DLEU1 deletions
  • Designed for use in chronic lymphocytic leukemia, multiple myeloma, and non-Hodgkin-lymphomas
  • Non-automated qualitative test method
  • Utilizes methanol/acetic-acid fixed cells from various sample types
  • Regulatory status: IVDR certified
  • 10 x 100 μL package configuration