Learn More
Radiometer America Radiometer FLEX PLUS ABL90 Solution Pack XL (980 Activities)
SDP
Catalog No. NC1702508
Encompass_Preferred
ABL90 FLEX PLUS Solution Pack XL (980 Activities)
980 activities Solution Pack for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
Catalog No. NC1702508 Supplier Radiometer America Supplier No. 944457
ABL90 FLEX PLUS Solution Pack XL (980 Activities)
980 activities Solution Pack for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.