Learn More
Keystone Safety XL-Sponbond Polypropylene Hood with Elastic Closure
SDP
Catalog No. 01257803
Change view
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-257-803 White
01-257-802 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-803 Supplier Keystone Safety Supplier No. HNWIECROWNXLWH
Add to Cart
Add to Cart
Made from breathable polypropylene with a secure elastic closure, it delivers comfortable protection while helping reduce contamination risks.
  • Head and hair coverage for cleanrooms, laboratories, and controlled environments
  • Extra-large size for improved coverage and comfort
  • Designed for users requiring additional fit and headroom (XL sizing)
  • Lightweight, breathable spunbond polypropylene material
  • Elastic closure for a secure fit
  • Helps reduce contamination risks from hair and particulate matter
  • Low-lint construction suitable for controlled environments
  • 100% latex-free
  • Disposable, single-use design
Specifications
Product Type Polypropylene Hood
Size X-Large
Material Spunbond Polypropylene
Color White
Closure Type Elastic
Disposable Single-use
Cleanroom Class Class 10,000 (ISO 7)